Learn More
Cayman Chemical Salvinorin A-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 791114-98-2, 1 mg
SDP
Catalog No. 502912421
Encompass_Preferred
An internal standard for the quantification of salvinorin A by GC- or LC-MS; intended for research and forensic applications
Catalog No. 50-291-2421 Supplier Cayman Chemical Supplier No. 364101 MG
An internal standard for the quantification of salvinorin A by GC- or LC-MS; intended for research and forensic applications