Learn More
Abcam Salvinorin A (Divinorin A), kappa agonist, 1MG
Catalog No. 502584705
Encompass_Preferred
MW 432.5 Da, Purity >98%. Potent κ agonist (Ki = 8-19 nM). Psychoactive and antinociceptive in vivo. Reported to be an allosteric modulator of μ opioid receptors.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4705 Supplier Abcam Supplier No. AB1200841MG
MW 432.5 Da, Purity >98%. Potent κ agonist (Ki = 8-19 nM). Psychoactive and antinociceptive in vivo. Reported to be an allosteric modulator of μ opioid receptors.
The product is subject to the following: Abcam Restricted Use Statement