Learn More
Cayman Chemical Salvinorin A Propionate, Purity: ≥98%, CAS: 689295-71-4, 10 mg
SDP
Catalog No. 502963984
Encompass_Preferred
A selective κ1-opioid receptor partial agonist (Ki = 32.6); inhibits forskolin-stimulated cAMP production (EC50 = 4.7 nM); antinociceptive effects in mice at 13 µg, i.c.v.
Catalog No. 50-296-3984 Supplier Cayman Chemical Supplier No. 2229010 MG
A selective κ1-opioid receptor partial agonist (Ki = 32.6); inhibits forskolin-stimulated cAMP production (EC50 = 4.7 nM); antinociceptive effects in mice at 13 µg, i.c.v.