Learn More
Abcam SAM e (S-(5-Adenosyl)-L-methionine disulfate tosylate), Methyl donor, 50MG
Numéro de catalogue. 502706275
Encompass_Preferred
MW 763.8 Da, Purity >95%. Methyl donor and cofactor for enzyme-catalyzed methylations, such as catechol O-methyltransferase (COMT) and DNA methyltransferases (DNMT).
The product is subject to the following: Abcam Restricted Use Statement
Numéro de catalogue. 50-270-6275 Fournisseur Abcam Code fournisseur AB14222150MG
MW 763.8 Da, Purity >95%. Methyl donor and cofactor for enzyme-catalyzed methylations, such as catechol O-methyltransferase (COMT) and DNA methyltransferases (DNMT).
The product is subject to the following: Abcam Restricted Use Statement