Learn More
Waters Corp Waters Reagent, SAM I, Normal Phase, Liquid, 20 mL
Catalog No. 50201706 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
SAM I Reagent is engineered for the separation of sugars and related compounds, utilizing Silica-Pak chromatographic cartridges. This reagent facilitates rapid separations of high sugar concentrations and maintains integrity against crude sample applications that can compromise other separation columns. Its design allows for mobile phase recirculation, ensuring efficient and economical use.
  • Separates sugars and related compounds such as Fructose, Glucose, Sucrose, Maltose, Lactose, Lactulose, Isomaltose, and Xylulose.
  • Formulated as a liquid with normal phase separation mode and RI detection.
  • Store at room temperature and ships ambient for convenience.
  • Compatible with LC system types.
  • Each package contains 5 bottles, each with 20 mL of reagent, sufficient to prepare 5 L of mobile phase or 0.5 L of pre-treatment solution.
Catalog No. 50-201-706 Supplier Waters Corp Supplier No. WAT010873
SAM I Reagent is engineered for the separation of sugars and related compounds, utilizing Silica-Pak chromatographic cartridges. This reagent facilitates rapid separations of high sugar concentrations and maintains integrity against crude sample applications that can compromise other separation columns. Its design allows for mobile phase recirculation, ensuring efficient and economical use.
  • Separates sugars and related compounds such as Fructose, Glucose, Sucrose, Maltose, Lactose, Lactulose, Isomaltose, and Xylulose.
  • Formulated as a liquid with normal phase separation mode and RI detection.
  • Store at room temperature and ships ambient for convenience.
  • Compatible with LC system types.
  • Each package contains 5 bottles, each with 20 mL of reagent, sufficient to prepare 5 L of mobile phase or 0.5 L of pre-treatment solution.