Learn More
American Radiolabeled Chemicals Inc SAM-METHYL-3H 1LC
SDP
Catalog No. 5090104350
Encompass_Preferred
Adenosyl-L-methionine, S-methyl-3H 1Lc in sulfuric acid (pH 2.0):ethanol (9:1), 1 mCi/ml, S.A. 55-85 Ci/mmol, M.W. 398.5, Shipped in Dry Ice, (SAM)
Catalog No. 50-901-04350 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 02881MC
Adenosyl-L-methionine, S-methyl-3H 1Lc in sulfuric acid (pH 2.0):ethanol (9:1), 1 mCi/ml, S.A. 55-85 Ci/mmol, M.W. 398.5, Shipped in Dry Ice, (SAM)