Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ SAM4 Plus Auscultation Manikin Light Skin
GREENER_CHOICESDPSCI_ED

Catalog No. S012486 Shop All 3B Scientific Products
Change view
Click to view available options
Light Skin
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S012486 Light Skin
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S012486 Supplier 3B Scientific™ Supplier No. 1025098
Add to Cart
Add to Cart
Request a Quote

The latest advancement in auscultation training from Cardionics.

  • Introducing SAM4, the latest advancement in auscultation training from Cardionics.
  • SAM4 builds upon the success of its predecessors, including SAM Basic, SAM II, and SAM 3G, and brings exclusive new features to elevate the auscultation learning experience.

Specifications

Product Type Auscultation Simulator
For Use With (Application) Teaching and Learning Auscultation
Color Light Skin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.