Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Samarium(II) Iodide Solution

Catalog No. 347116100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
4 x 25 mL
1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-711-6100ML 100 mL
34-711-625ML 25 mL
34-711-6 4 x 25 mL
34-711-61L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 34-711-6100ML Supplier Sigma Aldrich Supplier No. 347116100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Samarium(II) iodide solution (SmI2) can be used as a reagent in the synthesis of:Benzannulated pyrrolizidines and indolizidines by SmI2-induced cyclizations of indole derivatives.Chiral 4-substituted 2-oxazolidinones and 5,5-disubstituted oxazolidinones through asymmetric Reformatsky type reaction.+a-Aminoalkyl substituted +a-butyrolactones via ketyl-alkene coupling reaction.It can also be used in:Reduction of conjugated double and triple bonds into alkenes using SmI2/H2O/amine mixtures.Conversion of +a-hydroxyketones into 1,3-diols by SmI2/H2O/Et3N.Selective reduction of 6-membered lactones to the corresponding diols/triols using SmI2-H2O reagent system.SmI2 is an effective single-electron reducing agent for the promotion of ketone-olefin, ketyl aryl cyclizations, and pinacol coupling reactions under mild conditions. Often both intramolecular and intermolecular couplings proceed in a highly stereoselective fashion. It is also used in the synthesis of new heteroleptic samarium aryloxide/cyclopentadienide complexes.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 32248-43-4
Quantity 100 mL
Molecular Formula I2Sm
Linear Formula SmI2
MDL Number MFCD00058873
Molecular Weight (g/mol) 404.17 g/mol
Density 0.922 g/mL at 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.