Learn More
Strem, An Ascensus Company CAS# 13465-55-9. 25g. Samarium(III) chloride hexahydrate (99.9%-Sm) (REO). MFCD00149859
SDP
Catalog No. 5090116007
Encompass
CAS# 13465-55-9. 25g. Samarium(III) chloride hexahydrate (99.9%-Sm) (REO). MFCD00149859. Molecular Weight: 256.71 (364.80). Molecular Formula: SmCl3∙6H2O. Color/form: light yellow xtl. Strem# 93-6225. http://www.strem.com/catalog/v/93-6225/
Catalog No. 50-901-16007 Supplier Strem, An Ascensus Company Supplier No. 93622525G
CAS# 13465-55-9. 25g. Samarium(III) chloride hexahydrate (99.9%-Sm) (REO). MFCD00149859. Molecular Weight: 256.71 (364.80). Molecular Formula: SmCl3∙6H2O. Color/form: light yellow xtl. Strem# 93-6225. http://www.strem.com/catalog/v/93-6225/