Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Samarium(III) isopropoxide, 99% (REO), 5% w/v in toluene/isopropanol, Thermo Scientific™

Catalog No. AA41311AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41311AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA41311AE Supplier Thermo Scientific Chemicals Supplier No. 41311AE

missing translation for 'validMainframeMsg'

Precursor for CVD and Sol-Gel

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3504-40-3
Molecular Formula C9H24O3Sm
Molecular Weight (g/mol) 330.648
MDL Number MFCD00058820
InChI Key BVFXJLVXFKLRNG-UHFFFAOYSA-N
Synonym samarium iii isopropoxide
PubChem CID 85402317
IUPAC Name propan-2-ol;samarium
SMILES CC(C)O.CC(C)O.CC(C)O.[Sm]

Specifications

Odor Alcohol-like
Linear Formula Sm[OCH(CH3)2]3
Quantity 100 mL
UN Number UN3274
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 327.5
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 5% w/v in toluene/isopropanol
Physical Form Liquid
Assay (REO)
Chemical Name or Material Samarium(III) isopropoxide
Show More Show Less
Hazard Category H225-H304-H315-H319-H336-H361d-H373
Hazard Statement GHS H Statement
H225-H314-H318-H361-H373-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.