Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Samarium(III) nitrate hexahydrate

Catalog No. 298123100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-812-3100G 100 g
29-812-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-812-3100G Supplier Sigma Aldrich Supplier No. 298123100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Samarium(III) nitrate hexahydrate is a Lewis acid catalyst that is used in the synthesis of a variety of materials for applications such as:synthesis of nitrate precursor solution which can be used as a nanocatalyst in the solid oxide regenerative fuel cells.for the formation of coumarin derivative by von Pechmann condensation of phenol with ethyl acetoacetate in the presence of samarium complex as catalyst.for the preparation of samarium doped ceria, which can be used in the fabrication of electrolyte for fuel cells.Hydration may vary slightly

Specifications

CAS 13759-83-6
Quantity 100 g
Molecular Formula N3O9Sm -+ 6H2O
Linear Formula Sm(NO3)3 -+ 6H2O
MDL Number MFCD00149857
Synonym Samarium trinitrate; Trinitric acid samarium(III)-+6 hydrate
Molecular Weight (g/mol) 444.47
Percent Purity 99.9% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.