Learn More
Samarium(III) oxide, REacton™, 99.998% (metals basis), REM 100ppm
Description
Samarium(III) oxide is used in optical and infrared absorbing glass to absorb infrared radiation. It finds use as a neutron absorber in nuclear power reactors to control the reaction. It is also used as a catalyst for the dehydration of primary alcohols to aldehydes or ketones. It forms samarium perchlorate when treated with perchloric acid, which is used as a reference material for checking the wavelength scale of a spectrophotometer in the range of 200 to 500 nm.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 2300°C |
| Physical Form | Powder |
| Quantity | 100 g |
| Refractive Index | 1.97 |
| Merck Index | 14,8347 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 348.7 |
| Percent Purity | 99.998% |
| Grade | Reacton |
| Assay | (metals basis) |
| Show More |
Safety and Handling
| EINECSNumber | 235-043-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.