Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Samarium powder, <250 micron, REacton™, 99.9% (REO), Thermo Scientific™

Catalog No. AA1242206
Change view
Click to view available options
5g; Ampoule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AA1242206 5g; Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1242206 Supplier Thermo Scientific Chemicals Supplier No. 1242206

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-19-9
Molecular Formula Sm
MDL Number MFCD00011233
Quantity 5 g
UN Number UN3089
Synonym samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
InChI Key KZUNJOHGWZRPMI-UHFFFAOYSA-N
SMILES [Sm]
IUPAC Name samarium
Molecular Weight (g/mol) 150.36
PubChem CID 23951
ChEBI CHEBI:33374
Percent Purity 99.9%
Grade Reacton™
Odor Odorless
Packaging Ampule
Particle Size <250 Micron
Assay (REO)
Chemical Name or Material Samarium powder
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Precautionary Statement GHS P Statement
P210-P231+P232-P280-P370+P378b-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: Special powder for metal fires.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
TSCA TSCA
storageNote1
  • Ampouled under argon
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.