Learn More
Medchemexpress LLC Samatasvir | 1312547-19-5 | 99.39% | 885.06 | 100 MG
SDP
Catalog No. 5000393905
Encompass_Preferred
Samatasvir is a potent, orally active NS5A inhibitor of HCV replication. It is effective and selective against infectious HCV and replicons, with EC50s ranging from 2 to 24 pM in genotype 1 through 5 replicons. This compound maintains full activity even in the presence of HIV and hepatitis B virus (HBV) antivirals and does not exhibit cross-resistance with HCV protease, nucleotide, and nonnucleoside polymerase inhibitor classes.
  • Potent, orally active NS5A inhibitor of HCV replication.
  • Effective and selective against infectious HCV and replicons across genotypes 1 through 5.
  • Retains full activity with HIV and HBV antivirals.
  • No cross-resistance with other HCV inhibitor classes (protease, nucleotide, and nonnucleoside polymerase inhibitor classes).
Catalog No. 50-003-93905 Supplier Medchemexpress LLC Supplier No. HY16784100MG
Samatasvir is a potent, orally active NS5A inhibitor of HCV replication. It is effective and selective against infectious HCV and replicons, with EC50s ranging from 2 to 24 pM in genotype 1 through 5 replicons. This compound maintains full activity even in the presence of HIV and hepatitis B virus (HBV) antivirals and does not exhibit cross-resistance with HCV protease, nucleotide, and nonnucleoside polymerase inhibitor classes.
  • Potent, orally active NS5A inhibitor of HCV replication.
  • Effective and selective against infectious HCV and replicons across genotypes 1 through 5.
  • Retains full activity with HIV and HBV antivirals.
  • No cross-resistance with other HCV inhibitor classes (protease, nucleotide, and nonnucleoside polymerase inhibitor classes).