Learn More
Medchemexpress LLC Samatasvir | 1312547-19-5 | 99.4% | 885.06 | 50 MG
SDP
Catalog No. 5000393599
Encompass_Preferred
Samatasvir (IDX719) is a potent, orally active NS5A inhibitor that targets HCV replication. It demonstrates effectiveness and selectivity against infectious HCV and replicons, with EC50 values ranging from 2 to 24 pM across genotype 1 through 5 replicons.
  • Potent, orally active NS5A inhibitor of HCV replication.
  • Effective and selective against infectious HCV and replicons.
  • Maintains full activity in the presence of HIV and hepatitis B virus (HBV) antivirals.
  • Not cross-resistant with HCV protease, nucleotide, and nonnucleoside polymerase inhibitor classes.
Catalog No. 50-003-93599 Supplier Medchemexpress LLC Supplier No. HY1678450MG
Samatasvir (IDX719) is a potent, orally active NS5A inhibitor that targets HCV replication. It demonstrates effectiveness and selectivity against infectious HCV and replicons, with EC50 values ranging from 2 to 24 pM across genotype 1 through 5 replicons.
  • Potent, orally active NS5A inhibitor of HCV replication.
  • Effective and selective against infectious HCV and replicons.
  • Maintains full activity in the presence of HIV and hepatitis B virus (HBV) antivirals.
  • Not cross-resistant with HCV protease, nucleotide, and nonnucleoside polymerase inhibitor classes.