Learn More
Medchemexpress LLC Sambucus Nigra Lectin | ≥85.0% | 5 MG
SDP
Catalog No. 5000425722
Encompass_Preferred
Sambucus Nigra Lectin (SNA) is a plant lectin that can be used as a probe to specifically bind biomolecules such as polysaccharides and peptides. It is a biological material or organic compound suitable for life science research.
  • Can be used as a probe to specifically bind biomolecules (such as polysaccharides, peptides, etc.).
  • A biological material or organic compound suitable for life science research.
Catalog No. 50-004-25722 Supplier Medchemexpress LLC Supplier No. HYNP0805MG
Sambucus Nigra Lectin (SNA) is a plant lectin that can be used as a probe to specifically bind biomolecules such as polysaccharides and peptides. It is a biological material or organic compound suitable for life science research.
  • Can be used as a probe to specifically bind biomolecules (such as polysaccharides, peptides, etc.).
  • A biological material or organic compound suitable for life science research.