Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Samco™ Bio-Tite™ Specimen Containers for Sterile Environments
GREENER_CHOICE

Catalog No. 1371165 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
120 mL
Closure Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color
13-711-65 120 mL Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-711-65 Supplier Thermo Scientific™ Supplier No. 07 0006
  • $268.00
    $62.20 / Case of 100

    Save $205.80

    77% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Collect biopsy specimens and transport them into a sterile environment using Thermo Scientific™ Samco™ Bio-Tite™ Specimen Containers for Sterile Environments

Collect biopsy specimens and transport them into a sterile environment using Thermo Scientific™ Samco™ Bio-Tite™ Specimen Containers for Sterile Environments. These shatter-resistant containers are manufactured from virgin polypropylene and have a proprietary, all plastic, leak-resistant seal that prevents bio-hazardous spills. Bio-Tite containers are tested at varying pressures, making them ideal for use in various transport systems.

Features:
  • Dual-threaded caps for leak resistant seal
  • Sterile, individually wrapped, peel pouches for easy opening onto a sterile field
  • Available in sterile, economical baggy wrap

Ordering Information:

  • Case of 100
TRUSTED_SUSTAINABILITY

Specifications

Product Type Specimen Container
Closure Included Yes
Shape Cylindrical
Sterility Sterile
Capacity (Metric) 120 mL
Closure Color Blue
Closure Size 53 mm
Description Containers with caps
Diameter (Metric) 53 mm
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.