Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Samco™ Histology Bio-Tite™ Nonsterile Specimen Containers
GREENER_CHOICE

Catalog No. 1371194
Change view
Click to view available options
Capacity (Metric):
40 mL
120 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
13-711-94 40 mL
13-711-96 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-711-94 Supplier Thermo Scientific™ Supplier No. 6A 0010
Add to Cart
Edge
Add to Cart

Prevent bio-hazardous spills and conserve storage space with these stackable containers, offered in capacities from 20mL to 120mL with 35, 48 or 53mm openings.

Prevent bio-hazardous spills and conserve storage space with stackable Thermo Scientific™ Samco™ Histology Bio-Tite™ Nonsterile Specimen Containers, offered in capacities from 20mL to 120mL with 35, 48 or 53mm openings. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.

Features:
  • Dual-threaded caps create leak-resistant seal for superior safety and handling
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Virgin polypropylene allows higher clarity to see samples, and will not become brittle

Ordering Information:

Bulk packed, one bag per case. Caps must be ordered separately.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Specimen Container
Material Virgin Polypropylene
Shape Cylindrical
Sterility Non-sterile
Capacity (Metric) 40 mL
Closure Size 48 mm
Closure Type Bio-Tite Lid
Description 40mL/48mm Containers only
Diameter (Metric) 48 mm
Label No Label
No. per Case 600/Cs.
No. per Pack 600/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.