Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Samco™ Narrow Mouth Bio-Tite™ 40mL (1 oz.) 48mm Specimen Containers
GREENER_CHOICE

Catalog No. 1371186 Shop All Thermo Scientific Products
Change view
Click to view available options
Closure Color:
Yellow
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color
13-711-86 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-711-86 Supplier Thermo Scientific™ Supplier No. 04 0506
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prevent bio-hazardous spills and conserve storage space with these stackable containers, featuring a proprietary, all plastic, leak-resistant seal.

Prevent bio-hazardous spills and conserve storage space with stackable Thermo Scientific™ Samco™ Narrow-Mouth Bio-Tite™ 40mL (1 oz.) 48mm Specimen Containers. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.

Features:
  • Dual-threaded caps create leak-resistant seal for superior safety and handling
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Virgin polypropylene allows higher clarity to see samples, and will not become brittle
  • Sterile indicator tab assures tamper evidence
  • Bilingual English and Spanish label facilitates accurate usage

Ordering Information:

  • Available nonsterile or gamma irradiation sterilized
  • Available labled or non-labled
  • Available bulk or individually packed
  • In cases of 500
TRUSTED_SUSTAINABILITY

Specifications

Product Type Specimen Container
Closure Included Yes
Material Virgin Polypropylene
Shape Cylindrical
Sterility Non-sterile
Capacity (Metric) 40 mL
Closure Color Yellow
Closure Size 48 mm
Closure Type Bio-Tite Lid
Description Cap and container bagged separately
Diameter (Metric) 48 mm
Label Non-tabbed, 10% buffered formalin
No. per Case 600/Cs.
No. per Pack 600/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.