Learn More
Aviva Systems Biology Corp SAMD4A Antibody - middle region (ARP53707_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53707_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SAMD4A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SAMD4A. Alternate names: DKFZp434H0350, KIAA1053, SAMD4, SMG, SMGA, Smaug, Smaug1, SMAUG, SMAUG1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61102