Learn More
Aviva Systems Biology Corp SAMHD1 Antibody - middle region (ARP55262_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55262_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SAMHD1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human SAMHD1. Alternate names: AGS5, DCIP, HDDC1, MOP-5, SBBI88, CHBL2ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61448