Learn More
NEOGEN Colitag™ Sample Containers, Solid Plastic with Screw-top
SDP
Catalog No. 502015143
Encompass_Preferred
Approvals: EPA: Federal Register/vol. 74, No. 216/Tuesday, November 10, 2009/Rules and RegulationsModified Colitag™ is US EPA approved for compliance monitoring of public water systems as required by the Total Coliform Rule.Limit of Detection: 1 cfu in a 100 mL sampleFor more information visit https://www.neogen.com/solutions/Microbiology/Colitag-Sample-Containers
Catalog No. 50-201-5143 Supplier NEOGEN Supplier No. 700006475
Approvals: EPA: Federal Register/vol. 74, No. 216/Tuesday, November 10, 2009/Rules and RegulationsModified Colitag™ is US EPA approved for compliance monitoring of public water systems as required by the Total Coliform Rule.Limit of Detection: 1 cfu in a 100 mL sampleFor more information visit https://www.neogen.com/solutions/Microbiology/Colitag-Sample-Containers