Learn More
Perkin Elmer US LLC Polypropylene Disposable Sample Cups, 1.2 mL - Qty. 2000
Catalog No. 509052034
Encompass_Preferred
Sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample.
Catalog No. 50-905-2034 Supplier Perkin Elmer US LLC Supplier No. B0510397
Sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample.