Learn More
SKC Inc SAMPLE BAG,25L,DUAL S/S FITTNG
SDP
Catalog No. NC2181591
Encompass
Tedlar 25-liter Sample Bag with dual stainless steel fittings: the hose/valve for flushing, filling, and sealing the bag, and the septum with a syringe port for removing samples for analysis, pk/5External dimensions: 22 x 24 inches DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
This item is not returnable. View return policy
Catalog No. NC2181591 Supplier SKC Inc Supplier No. 23125
Tedlar 25-liter Sample Bag with dual stainless steel fittings: the hose/valve for flushing, filling, and sealing the bag, and the septum with a syringe port for removing samples for analysis, pk/5External dimensions: 22 x 24 inches DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”