Learn More
SKC Inc Tedlar Sample Bags, 1 Liter, Single Polypropylene Fitting
SDP
Catalog No. 50465244
Encompass
SKC Tedlar 1 Liter Sample Bag with single polypropylene septum fitting. Fitting combines the hose/valve and the septum holder into one compact fitting. This bag is suitable for use with Small Vac-U-Chamber (231-940), (pkg/10). DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”
Catalog No. 50-465-244 Supplier SKC Inc Supplier No. 23201
SKC Tedlar 1 Liter Sample Bag with single polypropylene septum fitting. Fitting combines the hose/valve and the septum holder into one compact fitting. This bag is suitable for use with Small Vac-U-Chamber (231-940), (pkg/10). DOES NOT ship to Alaska, Arizona, California, Colorado, Hawaii, Idaho, Nevada, Oregon, Utah, Washington. Wyoming”