Learn More
Electron Microscopy Sciences Glass Sample Bottle With Plastic Snap Cap, 8 ml
SDP
Catalog No. 501899144
Encompass_Preferred
For collecting and exhibiting a wide variety of specimens in the lab or field. Caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suitable for hydroscopic materials.
Capacity: 8 ml
Catalog No. 50-189-9144 Supplier Electron Microscopy Sciences Supplier No. 64252
For collecting and exhibiting a wide variety of specimens in the lab or field. Caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suitable for hydroscopic materials.
Capacity: 8 ml