Learn More
Hach Company Sample Bottle LDPE 30 mL
Supplier:  Hach Company 2087050
Encompass
Bottle-Sampling-Low Density-PE-30 mL-12/pkLow-density polyethylene (LDPE) bottles feature wide-mouth opening for easy filling and leak-proof polypropylene closure Great choice for grab samples shipping liquids soil sampling and storing dry chemicals
Catalog No. 50-002-07328