Learn More
Crescent Chemical Co Inc SAMPLE BUFFER FOR BLUE NATIVE
SDP
Catalog No. 50248596
Encompass_Preferred
Sample Buffer for Blue Native (2x) Sample buffer for Blue Native PAGE. Supplied as 2x concentrate.Contains 1 M 6-aminocaproic acid, 100 mM BisTris-HCl (pH 7.0), 100 mM NaCl, 20 % glycerol, 0.1 % SERVA Blue G. Size - 20ML Storage Conditions - -15 °C TO -25 °C Catalog
Catalog No. 50-248-596 Supplier Crescent Chemical Co Inc Supplier No. SE42533.01
Sample Buffer for Blue Native (2x) Sample buffer for Blue Native PAGE. Supplied as 2x concentrate.Contains 1 M 6-aminocaproic acid, 100 mM BisTris-HCl (pH 7.0), 100 mM NaCl, 20 % glycerol, 0.1 % SERVA Blue G. Size - 20ML Storage Conditions - -15 °C TO -25 °C Catalog