Learn More
Crescent Chemical Co Inc SAMPLE BUFFER FOR CLEAR NATIVE
SDP
Catalog No. 50248597
Encompass_Preferred
Sample Buffer for Clear Native (2x) Sample buffer for Clear Native PAGE. Supplied as 2x concentrate.Contains 100 mM NaCl, 100 mM imidazole, 4 mM 6-aminocaproic acid, 2 mM EDTA, 0.02 % Ponceau S, 20 % glycerol. Size - 20ML Storage Conditions - -15 °C TO -25 °C Catalog
Catalog No. 50-248-597 Supplier Crescent Chemical Co Inc Supplier No. SE42534.01
Sample Buffer for Clear Native (2x) Sample buffer for Clear Native PAGE. Supplied as 2x concentrate.Contains 100 mM NaCl, 100 mM imidazole, 4 mM 6-aminocaproic acid, 2 mM EDTA, 0.02 % Ponceau S, 20 % glycerol. Size - 20ML Storage Conditions - -15 °C TO -25 °C Catalog