Learn More
Perkin Elmer US LLC Sample Capillary for ST Nebulizers - 0.8 mm I.D., 700 μL/min, Blue
Catalog No. 502104926
Encompass_Preferred
This Sample Capillary, marked blue, has a self-aspiration rate of 700 µL/min at 1 L/min of Argon flow.
Catalog No. 50-210-4926 Supplier Perkin Elmer US LLC Supplier No. N8145137
This Sample Capillary, marked blue, has a self-aspiration rate of 700 µL/min at 1 L/min of Argon flow.