Learn More
Perkin Elmer US LLC Sample Capillary for ST Nebulizers - 0.5 mm I.D., 400 μL/min, Orange
Catalog No. 502104740
Encompass_Preferred
This Sample Capillary, marked orange, has a self-aspiration rate of 400 µL/min at 1 L/min of Argon flow.
Catalog No. 50-210-4740 Supplier Perkin Elmer US LLC Supplier No. N8122384
This Sample Capillary, marked orange, has a self-aspiration rate of 400 µL/min at 1 L/min of Argon flow.