Learn More
Hach Company Sample Cells for 2100A Turbidimeter
Supplier:  Hach Company 2100300
Encompass
Pack of six sample cells for use with 2100A Turbidimeter. 25x95 mm round glass, 30 mL.
Catalog No. 50-199-5381