Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach™ Sample Cells with Caps

Catalog No. 501995947 Shop All Hach Company Products
Encompass
Change view
Click to view available options
Capacity (Metric):
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
50-199-5947 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-5947 Supplier Hach™ Supplier No. 2410222
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Set of two square polystyrene cells with caps for spectroscopic measurement of liquids (25mL). Hach™ Sample Cells with Caps have an optical path of 1 in. for use in cases where absorption is low.

When the concentration of a sample is too low to be measured using a standard cell (optical path length of 10mm), a cell with a longer optical path length is used to enable measurement at the optimum absorption sensitivity.

TRUSTED_SUSTAINABILITY

Specifications

Type Sample cell
Color Clear with black cap
Capacity (Metric) 25 mL
Material Polystyrene
Includes Caps
Quantity 2/Pk.
Path Length 1 in.
For Use With (Application) Spectral Measurement of Liquids
Shape Square
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.