Learn More
Hach Company Sample Cells, 1 inch round, pk/6 (4 x 10 mL, 2 x 25 mL)
Supplier:  Hach Company 2838300
Encompass
Ideal for use with DR/2400 and DR/2500 spectrophotometers. Two 25 mL, 1 inch round glass sample cells with caps (10, 20, and 25 mL marks) and four 10 mL, 1 inch round glass sample cells with caps.
Catalog No. 50-199-6746