Learn More
Waters Corp Waters Sample Collection Plate, 384-well, Square Well, Polypropylene, 100 μL
Catalog No. 50781253 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This sample collection plate is designed for laboratory use, featuring a 384-well format with square wells and a 100 μL sample volume. Made from polypropylene, it offers excellent chemical resistance and meets SBS/ANSI compliance for robot compatible systems.
  • Made of polypropylene for chemical resistance
  • Features square wells with a 100 μL sample volume
  • Packaged in quantities of 50
  • Subjected to a strict quality control process to ensure no contaminants and meet high dimensional tolerance standards
  • Can serve as a collection plate for 96-well SPE and filtration-plate formats
Catalog No. 50-781-253 Supplier Waters Corp Supplier No. 186002631
This sample collection plate is designed for laboratory use, featuring a 384-well format with square wells and a 100 μL sample volume. Made from polypropylene, it offers excellent chemical resistance and meets SBS/ANSI compliance for robot compatible systems.
  • Made of polypropylene for chemical resistance
  • Features square wells with a 100 μL sample volume
  • Packaged in quantities of 50
  • Subjected to a strict quality control process to ensure no contaminants and meet high dimensional tolerance standards
  • Can serve as a collection plate for 96-well SPE and filtration-plate formats