Learn More
Waters Corp Waters Sample Collection Plate, 384-Well, Square, 250 μL, Polypropylene, SBS/ANSI Compliant
Catalog No. 50781254 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 384-well sample collection plate features square, 250 μL polypropylene wells, designed for the safe storage and safeguarding of numerous samples. It is SBS/ANSI compliant for robot-compatible systems and suitable for use with autosamplers. The polypropylene construction provides excellent chemical resistance, and the plate can also function as a collection plate for 96-well SPE and filtration-plate formats.
  • Chemically resistant polypropylene construction.
  • 384 square wells with 250 μL capacity.
  • SBS/ANSI compliant for robot-compatible systems.
  • Designed for use with autosamplers.
  • Can be used as a collection plate for 96-well SPE and filtration-plate formats.
  • Withstands up to 5000g of centrifugal force.
  • Protects samples and analytes from temperature changes and organic solvents when used with a sealant.
Catalog No. 50-781-254 Supplier Waters Corp Supplier No. 186002632
This 384-well sample collection plate features square, 250 μL polypropylene wells, designed for the safe storage and safeguarding of numerous samples. It is SBS/ANSI compliant for robot-compatible systems and suitable for use with autosamplers. The polypropylene construction provides excellent chemical resistance, and the plate can also function as a collection plate for 96-well SPE and filtration-plate formats.
  • Chemically resistant polypropylene construction.
  • 384 square wells with 250 μL capacity.
  • SBS/ANSI compliant for robot-compatible systems.
  • Designed for use with autosamplers.
  • Can be used as a collection plate for 96-well SPE and filtration-plate formats.
  • Withstands up to 5000g of centrifugal force.
  • Protects samples and analytes from temperature changes and organic solvents when used with a sealant.