Learn More
Waters Corp Waters 96-well Sample Collection Plate, Round Well, Polypropylene, 350 μL
Catalog No. 50781260 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 96-well sample collection plate, made of chemically resistant polypropylene, features round wells and a 350 μL well volume. It is designed for secure sample storage in laboratory settings and is compatible with robot systems and autosamplers, supporting applications like SPE and filtration-plate formats. It can withstand centrifugation up to 5000g.
  • SBS/ANSI compliant for robot-compatible systems.
  • Functions as a collection plate for 96-well SPE and filtration-plate formats.
  • Constructed from polypropylene for chemical resistance.
  • Designed to securely store and protect multiple samples.
  • Chemically inert.
  • Withstands centrifugation up to a maximum tolerance of 5000g.
  • When used with a sealant, protects samples and analytes from contamination, temperature fluctuation, and organic solvents.
Catalog No. 50-781-260 Supplier Waters Corp Supplier No. 186002643
This 96-well sample collection plate, made of chemically resistant polypropylene, features round wells and a 350 μL well volume. It is designed for secure sample storage in laboratory settings and is compatible with robot systems and autosamplers, supporting applications like SPE and filtration-plate formats. It can withstand centrifugation up to 5000g.
  • SBS/ANSI compliant for robot-compatible systems.
  • Functions as a collection plate for 96-well SPE and filtration-plate formats.
  • Constructed from polypropylene for chemical resistance.
  • Designed to securely store and protect multiple samples.
  • Chemically inert.
  • Withstands centrifugation up to a maximum tolerance of 5000g.
  • When used with a sealant, protects samples and analytes from contamination, temperature fluctuation, and organic solvents.