Learn More
Perkin Elmer US LLC Sample Compartment Iris Kit for the 150 mm Integrating Spheres
Catalog No. 502112895
Encompass_Preferred
The Reflectance Only Small Spot Kit is an optional accessory kit for the 150 mm Integrating Spheres module. The spheres module is compatible with LAMBDA 650/850/850+/950/1050/1050+ spectrometers.
Catalog No. 50-211-2895 Supplier Perkin Elmer US LLC Supplier No. L6020316
The Reflectance Only Small Spot Kit is an optional accessory kit for the 150 mm Integrating Spheres module. The spheres module is compatible with LAMBDA 650/850/850+/950/1050/1050+ spectrometers.