Learn More
Waters Corp Waters Corporation Sample Cone, 0.5 mm, Stainless Steel, MS System
Catalog No. 50466684 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sample Cone features a 0.5 mm aperture and is designed for use in MS system types.
  • Constructed from stainless steel.
  • Has an internal diameter of 0.5 mm.
  • Compatible with MS system types.
  • Compatible with SQ Detector 2 and Xevo TQD instruments.
  • Units per package: 1.
  • Replacement schedule as required.
Catalog No. 50-466-684 Supplier Waters Corp Supplier No. 700005997
This Sample Cone features a 0.5 mm aperture and is designed for use in MS system types.
  • Constructed from stainless steel.
  • Has an internal diameter of 0.5 mm.
  • Compatible with MS system types.
  • Compatible with SQ Detector 2 and Xevo TQD instruments.
  • Units per package: 1.
  • Replacement schedule as required.