Learn More
Waters Corp Waters Sample Cone, 0.8 mm, Stainless Steel, Mass Spectrometry
Catalog No. 50466646 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This replacement sample cone is designed for use with various mass spectrometry systems and is an integral part of the ion block assembly.
  • Designed for mass spectrometry applications.
  • Manufactured from stainless steel.
  • Features an internal diameter of 0.8 mm.
  • Includes one unit per package.
  • Recommended replacement schedule is as required.
  • Compatible with SYNAPT G2-S MS-HDMS, SYNAPT G2-Si MS-HDMS, SYNAPT G2-Si, MALDI SYNAPT G2-Si, Xevo TQS, and Synapt G2-S systems.
Catalog No. 50-466-646 Supplier Waters Corp Supplier No. 700005350
This replacement sample cone is designed for use with various mass spectrometry systems and is an integral part of the ion block assembly.
  • Designed for mass spectrometry applications.
  • Manufactured from stainless steel.
  • Features an internal diameter of 0.8 mm.
  • Includes one unit per package.
  • Recommended replacement schedule is as required.
  • Compatible with SYNAPT G2-S MS-HDMS, SYNAPT G2-Si MS-HDMS, SYNAPT G2-Si, MALDI SYNAPT G2-Si, Xevo TQS, and Synapt G2-S systems.