Learn More
Waters Corp Waters Corporation Sample Cone, Stainless Steel, 0.35 mm Internal Diameter, MS
Catalog No. 502039486 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Corporation Sample Cone is a spare part for Waters Mass Spectrometry (MS) systems. Constructed from stainless steel, it features a 0.35 mm internal diameter and is crucial for performance maintenance tasks related to the ion block in various MS instruments.
  • Constructed from stainless steel.
  • Features a 0.35 mm internal diameter.
  • Designed for Mass Spectrometry (MS) systems.
  • Included in performance maintenance kits for Xevo G2 QTof/Tof instruments.
  • Replaced during ion block maintenance.
Catalog No. 50-203-9486 Supplier Waters Corp Supplier No. 700004219
The Waters Corporation Sample Cone is a spare part for Waters Mass Spectrometry (MS) systems. Constructed from stainless steel, it features a 0.35 mm internal diameter and is crucial for performance maintenance tasks related to the ion block in various MS instruments.
  • Constructed from stainless steel.
  • Features a 0.35 mm internal diameter.
  • Designed for Mass Spectrometry (MS) systems.
  • Included in performance maintenance kits for Xevo G2 QTof/Tof instruments.
  • Replaced during ion block maintenance.