Learn More
Waters Corp Waters Sample Cone, 0.5 mm Internal Diameter, Stainless Steel
Catalog No. NC1646868 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sample Cone is a spare part designed for use in MS systems. It features a 0.5 mm internal diameter and is constructed from durable stainless steel, ensuring reliable performance in demanding laboratory environments.
  • Designed for MS systems.
  • Features a 0.5 mm internal diameter.
  • Constructed from stainless steel.
  • Supplied as 1 pk.
  • Replacement as required.
Catalog No. NC1646868 Supplier Waters Corp Supplier No. 700003055
This Sample Cone is a spare part designed for use in MS systems. It features a 0.5 mm internal diameter and is constructed from durable stainless steel, ensuring reliable performance in demanding laboratory environments.
  • Designed for MS systems.
  • Features a 0.5 mm internal diameter.
  • Constructed from stainless steel.
  • Supplied as 1 pk.
  • Replacement as required.