Learn More
Waters Corp Waters Sample Cone, 0.67 mm Aperture Diameter, Stainless Steel, MS System
Catalog No. 50466690 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This sample cone is designed for mass spectrometry (MS) systems. Crafted from durable stainless steel, it features a precise aperture diameter of 0.67 mm, ensuring optimal performance for sample introduction.
  • Designed for mass spectrometry (MS) systems.
  • Compatible with a wide range of mass spectrometry instruments.
  • Constructed from stainless steel.
  • Features an aperture diameter of 0.67 mm.
  • Recommended for replacement as required.
Catalog No. 50-466-690 Supplier Waters Corp Supplier No. 700009087
This sample cone is designed for mass spectrometry (MS) systems. Crafted from durable stainless steel, it features a precise aperture diameter of 0.67 mm, ensuring optimal performance for sample introduction.
  • Designed for mass spectrometry (MS) systems.
  • Compatible with a wide range of mass spectrometry instruments.
  • Constructed from stainless steel.
  • Features an aperture diameter of 0.67 mm.
  • Recommended for replacement as required.