Learn More
Waters Corp Waters Sample Cone Assembly, MS System, 1 pk
Catalog No. 502039817 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Sample Cone Assembly is a replacement component designed for Stepwave mass spectrometers. It plays a crucial role in the Ion Block Assembly, contributing significantly to the overall performance of the mass spectrometer.
  • Units per package: 1
  • System type: MS
  • Instrument compatibility: Xevo TQ Absolute, SELECT SERIES Cyclic IMS, SELECT SERIES MRT, SYNAPT XS, Xevo TQ-XS
  • Replacement schedule: As required
Catalog No. 50-203-9817 Supplier Waters Corp Supplier No. 700011217
This Sample Cone Assembly is a replacement component designed for Stepwave mass spectrometers. It plays a crucial role in the Ion Block Assembly, contributing significantly to the overall performance of the mass spectrometer.
  • Units per package: 1
  • System type: MS
  • Instrument compatibility: Xevo TQ Absolute, SELECT SERIES Cyclic IMS, SELECT SERIES MRT, SYNAPT XS, Xevo TQ-XS
  • Replacement schedule: As required