Learn More
Waters Corp Waters Sample Cone Assembly, Xevo TQ-S micro, Mass Spectrometer
Catalog No. 502039790 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Sample Cone Assembly is a replacement part for Stepwave mass spectrometers and is an integral component of the Ion Block Assembly, critical for the mass spectrometer's performance. This specific Sample Cone Assembly is designed for the Xevo TQ-S micro mass spectrometers.
  • Replacement part for Stepwave mass spectrometers
  • Integral component of the ion block assembly
  • Critical for mass spectrometer performance
  • One unit per package
  • Sample cone type
  • MS system type
  • Replacement is as required
Catalog No. 50-203-9790 Supplier Waters Corp Supplier No. 700010289
The Sample Cone Assembly is a replacement part for Stepwave mass spectrometers and is an integral component of the Ion Block Assembly, critical for the mass spectrometer's performance. This specific Sample Cone Assembly is designed for the Xevo TQ-S micro mass spectrometers.
  • Replacement part for Stepwave mass spectrometers
  • Integral component of the ion block assembly
  • Critical for mass spectrometer performance
  • One unit per package
  • Sample cone type
  • MS system type
  • Replacement is as required