Learn More
Thermo Scientific Sample and Skimmer Cones | Nickel | 1 EA
Catalog No. NC0721740 Shop All Thermo Scientific Products
Encompass
Easily changeable, Designed for durability, performance, and ease of use, High-quality field-proven cones, Reliable high-sensitivity analysis down to detection-limit level by ICP-MS, Available in solid Ni construction, Pt-tipped for even greater durability, Al for specific analysis requirements
Catalog No. NC0721740 Supplier Thermo Scientific Supplier No. 1260630
Easily changeable, Designed for durability, performance, and ease of use, High-quality field-proven cones, Reliable high-sensitivity analysis down to detection-limit level by ICP-MS, Available in solid Ni construction, Pt-tipped for even greater durability, Al for specific analysis requirements