Learn More
Hach Company SAMPLE CONTAINER, 120ML HIGH PF
Supplier:  Hach Company 2552200
Encompass
These 120mL (4oz.) High Profile Corning Snap-Seal™ polypropylene containers are designed for a wide variety of applications and provide a reliable leak-tight seal when closed properly. Approximately 3.5 inches tall (91 mm) and 1.75 inches (45 mm) diameter. Plastic, snap seal, translucent. Sold individually.
Catalog No. 50-199-6220