Learn More
Hach Company Sample Container, 300 mL
Supplier:  Hach Company 2552511
Encompass
These 300mL (10oz.) Corning Snap-Seal™ polypropylene containers are designed for a wide variety of applications and provide a reliable leak-tight seal when closed properly. Approximately 4.4 inches tall (112 mm) and 2.48 inches (63 mm) diameter. Plastic, snap seal, translucent. 100/pk
Catalog No. 50-199-6223