Learn More
Hach Company Sample Container, polypropylene, 45 mL
Supplier:  Hach Company 2552114
Encompass
These 45mL (1.5oz.) Corning Snap-Seal™ polypropylene containers are designed for a wide variety of applications and provide a reliable leak-tight seal when closed properly. Approximately 3.3 inches tall (84 mm) and 1.18 inches (30 mm) diameter. Plastic, snap seal, translucent. 600/pk
Catalog No. 50-199-6219