Learn More
Hach Company Sample Container, 120 mL, PK. 250 Containers
Supplier:  Hach Company 2552212
Encompass
These 120mL (4oz.) High Profile Corning Snap-Seal™ polypropylene containers are designed for a wide variety of applications and provide a reliable leak-tight seal when closed properly. Approximately 3.5 inches tall (91 mm) and 1.75 inches (45 mm) diameter. Plastic, snap seal, translucent. 250/pk
Catalog No. 50-199-6221